ContactUsLinkComponent
Jalas Rigger Safety 1868 King, Wide fit with Aluminium Toecap & Steel Midsole, PU-Coated Leather Upper, PU Pronose, Nitrile Rubber O utsole, Thinsulate Heat Insulation , HR Upper, Oil-resistant outsole, water repellant, anti-static properties, ESD, d 1 * 1 PAIR
Catalog # EJEN1868W-47
Supplier:  EJENDALS
undefined
Jalas Rigger Safety 1868 King, Wide fit with Aluminium Toecap & Steel Midsole, PU-Coated Leather Upper, PU Pronose, Nitrile Rubber O utsole, Thinsulate Heat Insulation , HR Upper, Oil-resistant outsole, water repellant, anti-static properties, ESD, d 1 * 1 PAIR
Catalog # EJEN1868W-47
Supplier:  EJENDALS