Biological Indicatorological Indicator Spore Coupons for Monitoring Hydrogen Peroxide / Plasma Processes, Bacillus atrophaeus Cell L ine 9372, Stainless Steel Coupons each 57 x 70 mm Individually Packaged in Tyvek®/Mylar® Pouches, 100 per box, 104 p 1 * 1 Each
Catalog # NAMSCPN-04
Supplier: CROSSTEX
Frequently Bought Together