ContactUsLinkComponent
Mini Self-Contained Biological Indicatorological Indicator (SCBiological Indicators) for Monitoring Ethylene Oxide (EO) Sterilizati on Processes, Bacillus atrophaeus Cell Line 9372, SCBiological Indicators each 9 mm x 50 mm, 106 population level, 10 1 * 1 Each
Catalog # NAMSSCA-100
Supplier:  CROSSTEX
Mini Self-Contained Biological Indicatorological Indicator (SCBiological Indicators) for Monitoring Ethylene Oxide (EO) Sterilizati on Processes, Bacillus atrophaeus Cell Line 9372, SCBiological Indicators each 9 mm x 50 mm, 106 population level, 10 1 * 1 Each
Catalog # NAMSSCA-100
Supplier:  CROSSTEX
Frequently Bought Together