ContactUsLinkComponent
Spore Wire Inoculated Carriers for Monitoring Ethylene Oxide (EO) Processes, Bacillus atrophaeus Cell Line 9372, Braided Wires each 40 mm in length and <2 mm in Diameter, Packaged in Bulk, 100 per box, 106 population level, ISO 11138 and USP complian 1 * 1 Each
  NAMSSWN-06
 :  CROSSTEX
Spore Wire Inoculated Carriers for Monitoring Ethylene Oxide (EO) Processes, Bacillus atrophaeus Cell Line 9372, Braided Wires each 40 mm in length and <2 mm in Diameter, Packaged in Bulk, 100 per box, 106 population level, ISO 11138 and USP complian 1 * 1 Each
  NAMSSWN-06
 :  CROSSTEX
Frequently Bought Together