ContactUsLinkComponent
Biological Indicatorological Indicator Mini-Spore Strips for Monitoring Ethylene Oxide (EO) or Dry Heat Processes, Bacillus atrophae us Cell Line 9372, 2 mm x 10 mm Strips Individually Packaged in 25 mm x 25 mm Mini-Glassine Envelopes, 100 strips per 1 * 1 Each
Catalog # NAMSSTN-062MG
Supplier:  CROSSTEX
Biological Indicatorological Indicator Mini-Spore Strips for Monitoring Ethylene Oxide (EO) or Dry Heat Processes, Bacillus atrophae us Cell Line 9372, 2 mm x 10 mm Strips Individually Packaged in 25 mm x 25 mm Mini-Glassine Envelopes, 100 strips per 1 * 1 Each
Catalog # NAMSSTN-062MG
Supplier:  CROSSTEX
Frequently Bought Together