ContactUsLinkComponent
Spore Suspension, Bacillus subtilis Cell Line 5230, 10 mL, 104 population level per 0.1 mL, ISO 11138 and USP compliant where appli cable 1 * 10 mL
Catalog # NAMSUS52304
Supplier:  CROSSTEX
Spore Suspension, Bacillus subtilis Cell Line 5230, 10 mL, 104 population level per 0.1 mL, ISO 11138 and USP compliant where appli cable 1 * 10 mL
Catalog # NAMSUS52304
Supplier:  CROSSTEX
Frequently Bought Together