VWR®, Storage Boxes
180×120×75 mm
Artikelnr. 216-4000
Anbieter: VWR Collection
Jump to:
Product Details & Documents
These boxes are an ideal choice for laboratories due to their high transparency and excellent durability.
- Stackable, transparent boxes
- For storage and presentation
- Six different sizes
- PS, clear
Dokumente
Specifications
- W×D×H:180×120×75 mm