ContactUsLinkComponent
Super Streptavidin (SSA) Biosensors - Five trays of 96 biosensors coated with streptavidin for small molecule kinetic analysis. 10 % Discount on tray pricing included. 1 * 5 items
  PALF18-5065
Super Streptavidin (SSA) Biosensors - Five trays of 96 biosensors coated with streptavidin for small molecule kinetic analysis. 10 % Discount on tray pricing included. 1 * 5 items
  PALF18-5065
Frequently Bought Together