NIST TRACEABLE MONODISPERSE PARTICLE STANDARD 9.18 µM + COFA 1 * 1 items
Artikelnr. WHITMS0009
Supplier: WHITEHOUSE
NIST TRACEABLE MONODISPERSE PARTICLE STANDARD 9.18 µM + COFA 1 * 1 items
Artikelnr. WHITMS0009
Supplier: WHITEHOUSE
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Turpentine oil
Supplier: Roth Carl
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
GLYCEROL PURISS., MEETS ANALYTICAL SPECI 1 * 1 L
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
NIST TRACEABLE MONODISPERSE PARTICLE STANDARD25.60 µM + COFA 1 * 1 items
Supplier: WHITEHOUSE
2-Methylbutane for HPLC
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals