LigaTrap resins bind an exceptionally broad range of recombinant, monoclonal, and human/animal source polyclonal immunoglobulins. L igaTrap resins provide distinct advantages to traditional bind and elute purifications resins including mild elution 1 * 5 mL
Catalog # LIGALT-136
Supplier: LIGATRAP TECHNOLOGIES
Frequently Bought Together