ContactUsLinkComponent
ARTHUR CALIBRATION BEADS Alignment beads for Arthur fluorescence cell counter. Contains: Green calibration beads and red calibration beads (100µl each). 1 * 1 KIT
Catalog # NANOAB0100
Supplier:  NANOENTEK
ARTHUR CALIBRATION BEADS Alignment beads for Arthur fluorescence cell counter. Contains: Green calibration beads and red calibration beads (100µl each). 1 * 1 KIT
Catalog # NANOAB0100
Supplier:  NANOENTEK
Frequently Bought Together