Order Entry
Austria
Orders LinkContactUsLinkComponent
Electrolyte solutions

Electrolyte solutions

Electrolyte solution (KCl 3M+AgCl), 250 ml, 250 mL
Catalog # HACHLZW9501.99
Supplier:   Hach
Electrolyte solutions
Catalog # HACHLZW9501.99
Supplier:  Hach
CAS Number:  
Electrolyte solutions
Electrolyte solutions

Electrolyte solution (KCl 3M+AgCl), 250 ml, 250 mL

CAS Number:  
Price
28,80 €
Log in for availability
1 * 250 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Electrolyte solution (KCl 3M+AgCl), 250 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.