Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions, colour coded

pH buffer solutions, colour coded

Buffer solution, 4,01, ±0,02, Phthalate, Red, 25 °C, 15 mL
Catalog # HACH2283436
Supplier:   Hach
pH buffer solutions, colour coded
Catalog # HACH2283436
Supplier:  Hach
CAS Number:  
pH buffer solutions, colour coded
pH buffer solutions, colour coded

Buffer solution, 4,01, ±0,02, Phthalate, Red, 25 °C, 15 mL

CAS Number:  
Price
9,50 €
Log in for availability
1 * 15 mL
Product Details & Documents

For calibration of pH meters

  • Colour coded for easy identification
  • Accuracy (pH): ±0,02 at 25 °C

Certifications: Traceable to NIST.

Specifications

Specifications

  • pH buffer type:
    Phthalate
  • Pk:
    15 mL
  • Reference temperature:
    25 °C
  • Colour:
    Red
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,02
  • pH value:
    4,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.