XTip 250 µL Reload System Pipet Tips. Clean Pack Reloads, Low Retention, Pre-sterile, Rack Lid and Base, 96 Tip/Card, 10 Cards/Pack, 5 Pack/Case. Patented features: FlexFit™ for superior seal, X-Resin™ for low sample retention, Blade™ for accuracy, b 1 * 960 items
Catalog # 613-5458
Supplier: BIOTIX
Frequently Bought Together