ContactUsLinkComponent
This product is prepared by labeling highly cross-adsorbed goat anti-rabbit IgG (H+L) with CF430. To minimize cross-reactivity, the antibody has been adsorbed against human, mouse, and rat serum. 1 * 1 mg
Catalog # 20460-1MG
Supplier:  Biotium
This product is prepared by labeling highly cross-adsorbed goat anti-rabbit IgG (H+L) with CF430. To minimize cross-reactivity, the antibody has been adsorbed against human, mouse, and rat serum. 1 * 1 mg
Catalog # 20460-1MG
Supplier:  Biotium
Frequently Bought Together