ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RABBIT IgG (WHOLE MOLECULE). Anti-Rabbit Host: goat Form: affinity purifi ed liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855689
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RABBIT IgG (WHOLE MOLECULE). Anti-Rabbit Host: goat Form: affinity purifi ed liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855689
Supplier:  MP Biomedicals
Frequently Bought Together