ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO RABBIT IgG Fc. Anti-Rabbit Host: goat Form: lyophilized (IgG) Applications: Immunostain ing of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855679
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO RABBIT IgG Fc. Anti-Rabbit Host: goat Form: lyophilized (IgG) Applications: Immunostain ing of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855679
Supplier:  MP Biomedicals
Frequently Bought Together