ContactUsLinkComponent
MP SILICA GF-TLC. Contains 11% GypsumMean Pore Size: 60ÅSpecific Surface Area: 500-600 m2 /gParticle Size: 5-15 µmWeight: 0.4 g/mlpH : 6.5-7.0 1 * 500 g
Catalog # ICNA0204645
Supplier:  MP Biomedicals
MP SILICA GF-TLC. Contains 11% GypsumMean Pore Size: 60ÅSpecific Surface Area: 500-600 m2 /gParticle Size: 5-15 µmWeight: 0.4 g/mlpH : 6.5-7.0 1 * 500 g
Catalog # ICNA0204645
Supplier:  MP Biomedicals
Frequently Bought Together