ContactUsLinkComponent
AGAR BACTERIOLOGICAL GRADE. Molecular Biology ReagentSpecially purified for use in preparing solid culture media for microbiological and bacteriological applications. Naturally occurring impurities have been reduced to a minimum. 1 * 250 g
Catalog # ICNA0219402183
Supplier:  MP Biomedicals
AGAR BACTERIOLOGICAL GRADE. Molecular Biology ReagentSpecially purified for use in preparing solid culture media for microbiological and bacteriological applications. Naturally occurring impurities have been reduced to a minimum. 1 * 250 g
Catalog # ICNA0219402183
Supplier:  MP Biomedicals
Frequently Bought Together