AC 200-S49 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC200-S49, Amb+13 to 200 C, 28-49 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) & LONG SHAFT REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 1 * 1 items
Artikelnr. HAKA1561491
Supplier: THERMO ELECTRON LED
Frequently Bought Together