ContactUsLinkComponent
AC 200-S21 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC200-S21, Amb+13 to 200 C, 7-17 L VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 8 LANGUAGES 1 * 1 items
Catalog # HAKA1561211
AC 200-S21 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC200-S21, Amb+13 to 200 C, 7-17 L VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 8 LANGUAGES 1 * 1 items
Catalog # HAKA1561211
Frequently Bought Together