Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Rectangular cuvettes

Rectangular cell with lid, 45 mm, 200 µl, 1 mm, Optical glass
  SCLI80-2003-83
 :   BIOCHROM
Rectangular cuvettes
  SCLI80-2003-83
 :  BIOCHROM
Rectangular cuvettes

Rectangular cell with lid, 45 mm, 200 µl, 1 mm, Optical glass

 
 
   
1 * 1 ST
 
 
  • Path length:
    1 mm
  • Volume:
    200 µl
  • Window material:
    Optical glass
  • Description:
    Rectangular cell with lid
  • Height:
    45 mm
  • Seq:
    02