Microscope slides for printers
Orange
Catalog # 631-1236
Supplier: KLINIPATH
Jump to:
Product Details & Documents
Microscope slides for slide printers allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
- Frosted ends
- 90° grounded edges
- Finished clipped corners of 1mm
Specifications
- Colour:Orange
- Seq:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



