ContactUsLinkComponent
Cleanroom SpillEx gamma-irradiated - For sterile areas - Spill pick-up pad for all water-based liquids (drug manufacturing, body fluids…) - Not suitable for hazardous chemicals 1 * 50 items
Catalog # VILE150739
Supplier:  FHP VILEDA SCS
Cleanroom SpillEx gamma-irradiated - For sterile areas - Spill pick-up pad for all water-based liquids (drug manufacturing, body fluids…) - Not suitable for hazardous chemicals 1 * 50 items
Catalog # VILE150739
Supplier:  FHP VILEDA SCS
Frequently Bought Together