Orders LinkContactUsLinkComponent
Single-use bags for HYPERStack® vessels
Quench bag, 5 L
  MDTC91-200-76
 :   Corning
Single-use bags for HYPERStack® vessels
  MDTC91-200-76
 :  Corning
Single-use bags for HYPERStack® vessels
Quench bag, 5 L
211,89 €
1 * 1 ST
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
  • Capacity:
    5 L
  • Description:
    Quench bag
  • Seq:
    02