Vanillin NF
50 kg
Artikelnr. SPCMVA130-50KGBL
Anbieter: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: 4-Hydroxy-3-methoxybenzaldehyde
- CAS Number:121-33-5
- Formula:C₈H₈O₃
- MDL Number:MFCD00006942
- Molecular Weight:152.15 g/mol
- Merck Index:13,09998
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:∼82 °C
- Density:1.056 g/cm³ (20 °C)
- Flash Pt:153 °C
- Storage Temperature:Ambient
- EINECS:204-465-2
Specifications
Specification Test Results
- Assay:97% Min.
Specifications
- Pk:50 kg



