Order Entry
Austria
Orders LinkContactUsLinkComponent
Fumaric acid NF

Fumaric acid NF

Poly bottle, 2,5 kg
Catalog # SPCMF1096-2.5KG
Irritant
Supplier:   Spectrum Chemical
Fumaric acid NF
Catalog # SPCMF1096-2.5KG
Supplier:  Spectrum Chemical
CAS Number:  110-17-8
Fumaric acid NF

Poly bottle, 2,5 kg

CAS Number:  110-17-8
Price
1 660,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116.07 g/mol
  • Merck Index:13,04308
  • Boiling Pt:356 °C (1013 hPa)
  • Melting Pt:287 °C
  • Density:1.625 g/cm³ (20 °C)
  • Flash Pt:273 °C
  • EINECS:203-743-0
Specifications

Specification Test Results

  • Assay:
    99.5% Min.

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.