Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetanilide

Acetanilide

Poly pail, 12 kg
Catalog # SPCMA1007-12KGBL
Irritant
Supplier:   Spectrum Chemical
Acetanilide
Catalog # SPCMA1007-12KGBL
Supplier:  Spectrum Chemical
CAS Number:  103-84-4
Acetanilide

Poly pail, 12 kg

CAS Number:  103-84-4
Price
1 470,00 €
Log in for availability
1 * 12 kg
Product Details & Documents
Synonyms:  N-Phenylacetamide
  • CAS Number:103-84-4
  • Formula:C₈H₉NO
  • MDL Number:MFCD00008674
  • Molecular Weight:135.17 g/mol
  • Merck Index:13,00051
  • Boiling Pt:304 °C (1013 hPa)
  • Melting Pt:115 °C
  • Density:1.22 g/cm³ (20 °C)
  • Flash Pt:174 °C
  • EINECS:203-150-7
Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg

Specifications