Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER PH 7,00 +/-0,01 @20C (BAG IN BOX) 1 * 5 L
Catalog # REGEBPH22
Supplier:
REAGECON
Expand image
BUFFER PH 7,00 +/-0,01 @20C (BAG IN BOX) 1 * 5 L
Catalog # REGEBPH22
Supplier:
REAGECON
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Hydrochloric acid 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo