Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Unisette™, M405

Tissue processing/embedding cassettes, Unisette™, M405

Cream
Catalog # SIMPM405-8
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M405
Catalog # SIMPM405-8
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M405
Tissue processing/embedding cassettes, Unisette™, M405

Cream

Price
231,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

These cassettes are moulded from a special high density polymer and keep specimens safely submerged in liquid.

  • Resistant to the chemical action of most histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Snap latch and hinge-lock design prevent early separation of base and lid

Lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss.

Ordering information: Available in non-cytotoxic, non-metallic colors.

Packaging: Available in 25 stacks of 40 cassettes or 3 dispenser boxes of 500 cassettes.

Specifications
  • Colour:
    Cream
  • Seq:
    07