ContactUsLinkComponent
PV-1 PERFORMANCE VERIFICATION SOLUTION 1 Ampule à 0.5 ml For wavelength and photometric performance verification of the NanoDrop ONE 1 * 0,5 mL
  NANDND-PV-1
PV-1 PERFORMANCE VERIFICATION SOLUTION 1 Ampule à 0.5 ml For wavelength and photometric performance verification of the NanoDrop ONE 1 * 0,5 mL
  NANDND-PV-1
Frequently Bought Together