Orders LinkContactUsLinkComponent
Test charts
Test charts
Opacity scrub charts with optical brightner, White/black, 165×480 mm
Catalog # SIMEVF2316
Supplier:   Industrial Physics
Test charts
Catalog # SIMEVF2316
Supplier:  Industrial Physics
Test charts
Test charts
Opacity scrub charts with optical brightner, White/black, 165×480 mm
Price
285,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
  • Dimensions from A6 up to A4
  • Film laminated for an excellent solvent, chemical resistance, and film spread
  • Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Ordering information: A certificate is available for charts with an optical brightener.
Specifications
  • Colour:
    White/black
  • Description:
    Opacity scrub charts with optical brightner
  • Seq:
    12
  • W×H:
    165×480 mm