Express Eingabe
Austria
AuftragsverfolgungKontakt
Permeability cups
Permeability cups
Permeability cup with a surface area of 25 cm²
Catalog # SIMEVF2201
Supplier:   Industrial Physics
Permeability cups
Catalog # SIMEVF2201
Supplier:  Industrial Physics
Permeability cups
Permeability cups
Permeability cup with a surface area of 25 cm²
Price
278,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Permeability cups are designed to determine the water vapour transmission of paints, varnishes, coatings and related products. Permeability cup is suitable for testing both self-supporting coating and non-self-supporting coatings.
  • Anodised aluminium for corrosion resistance and colouring
  • Seal ring to prevent turning when closing the cover
  • Any deformation to the cup may cause deviation in the result
The wet cup method is the reference for determining water vapour transmission.
 : ISO 7783 (supersedes NF T30-018), ASTM D1653, ASTM E95.
 : Includes a cup, a seal ring and cover ring.
Documents
Specifications
  • Bezeichnung:
    Permeability cup with a surface area of 25 cm²
  • Seq:
    02