ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine, horseradish peroxidase (AS15 TMB-HRP-1L) is a sensitive reagent that is ready to use for the quantit ative detection of HRP bound to a solid phase or in free solution. 1 * 1 L
Catalog # AGRIAS15TMB-HRP-1L
Supplier:  Agrisera
3,3',5,5'-Tetramethylbenzidine, horseradish peroxidase (AS15 TMB-HRP-1L) is a sensitive reagent that is ready to use for the quantit ative detection of HRP bound to a solid phase or in free solution. 1 * 1 L
Catalog # AGRIAS15TMB-HRP-1L
Supplier:  Agrisera