ContactUsLinkComponent
Streptavidin is a protein that has a very high affinity for biotin (known as vitamin B7 or H). The binding of biotin to streptavidin is one of the strongest non-covalent interactions known in nature.temperature and pH. 1 * 1 mg
Catalog # AGRIAS152860
Supplier:  Agrisera
Streptavidin is a protein that has a very high affinity for biotin (known as vitamin B7 or H). The binding of biotin to streptavidin is one of the strongest non-covalent interactions known in nature.temperature and pH. 1 * 1 mg
Catalog # AGRIAS152860
Supplier:  Agrisera