BW CLIP CO 0-300PPM 30/60 3 YEARS 1 * 1 items
Catalog # BWTEBWC3-M3060
Supplier: BW TECHNOLOGIES
Frequently Bought Together
Camera adapters
Supplier: MOTIC
pH/mV/°C meters, handheld, FiveGo™ F2
Supplier: Mettler - Toledo
Filter funnels, VitraPOR®
Supplier: ROBU GLASFILTER
VWR®, Combs for PerfectBlue Gel System Mini L
Supplier: VWR Collection
LED LAMPE 3X2W DAYLIGHT 6000K GU5,3 1 * 1 items
Supplier: ZEISS
Barbed luer adapters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
Pocket refractometer, Brix/RI-Chek
Supplier: REICHERT
VWR®, Accessories for PerfectBlue Sedec Semi-Dry Blotters
Supplier: VWR Collection
Vials 1 * 1 items
Supplier: GILSON LCMS
Titrator, Titronic® 300
Supplier: SI Analytics
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Supplier: VWR Collection
Storage phosphor screens and cassettes
Supplier: Cytiva
VWR®, Combs for PerfectBlue™ Wide Format Gel System Midi ExW
Supplier: VWR Collection
VWR®, Cell Holders
Supplier: VWR Collection