Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Pre-mixed, 1000 µg/ml Pd and 600 µg/ml Mg(NO₃)₂, 250 ml, 2% HNO₃, 250 mL
Catalog # PROOVHG-MPM2-250
Supplier: LGC Standards PROMOCHEM
Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Catalog # PROOVHG-MPM2-250
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
This solution is intended for use as a matrix modifier for graphite furnace atomic absorption spectroscopy (GFAA) in the determination of trace elements.
Certifications: Traceable to NIST, SRM.
Specifications
Specifications
- Matrix:2% HNO₃
- Pk:250 mL
- Description:Pre-mixed, 1000 µg/ml Pd and 600 µg/ml Mg(NO₃)₂, 250 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



