Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
1% Magnesium nitrate, 100 ml, 2% HNO₃, 100 mL
Catalog # PROOVHG-MMGN1P-100
Supplier: LGC Standards PROMOCHEM
Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Catalog # PROOVHG-MMGN1P-100
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
This solution is intended for use as a matrix modifier for graphite furnace atomic absorption spectroscopy (GFAA) in the determination of trace elements.
: Traceable to NIST, SRM.
Specifications
Specifications
- Matrix:2% HNO₃
- Pk:100 mL
- Description:1% Magnesium nitrate, 100 ml



