Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TEMAZEPAM IMPURITY C (3RS)-7-CHLORO-1-METHYL-2-OXO-5-PHENYL-2,3-DIHYDRO-1H-1,4-BENZODIAZEPIN-3-YL ACETATE (TEMAZEPAMACETAT) 1 * 5 mg
Catalog # EDQMY0000344
Supplier:
EDQM
Expand image
TEMAZEPAM IMPURITY C (3RS)-7-CHLORO-1-METHYL-2-OXO-5-PHENYL-2,3-DIHYDRO-1H-1,4-BENZODIAZEPIN-3-YL ACETATE (TEMAZEPAMACETAT) 1 * 5 mg
Catalog # EDQMY0000344
Supplier:
EDQM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals