GLYCOPYRRONIUM FOR PEAK IDENTIFICATION 1 * 10 mg
Catalog # EDQMY0001309
Supplier: EDQM
Frequently Bought Together
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1078302
Supplier: VWR Chemicals
Lithium standard for IC, TraceCERT®, Supelco®
Supplier: Merck
ICP METAL STANDARD 31 COMPONENT 1 * 100 mL
Supplier: CPACHEM