PRAZIQUANTEL FOR SYSTEM SUITABILITY 1 * 10 mg
Artikelnr. EDQMY0001533
Supplier: EDQM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Organic reference standard, Benzotrifluoride 2000 µg/ml in methanol, Dr Ehrenstorfer
Supplier: EHRENSTORFER
PHOSPHONIC ACID 100µG/ML IN ACETONITRILE 1 * 1 mL
Supplier: EHRENSTORFER
Dienochlor certified 1 * 0,25 g
Supplier: EHRENSTORFER
Stands for SCOTTY® cylinders
Supplier: Merck
Organic reference standard, spinetoram, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Betamethasone Valerate, Reference Standards
Supplier: USP
Supelco® Organic Reference Standard, (+)-Menthofuran
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Dessiccant, Reference Standards
Supplier: USP
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
METOPROLOL EP IMPURITY G 1 * 50 mg
Supplier: Merck
Pirimicarb Analytical standard, reference material, Fluka™
Supplier: Honeywell Chemicals
ARCTIIN, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck