Rotavapor R-300 no bath, SJ29/32, no glass, I-300, VU, 230V. . Lift System: Elec. Heating Bath: w/o SJ 29/32 Glass set: w/o int.: I -300, valve unit 1 * 1 items
Catalog # BUCH11R300201N041
Supplier: BUCHI
Rotavapor R-300 no bath, SJ29/32, no glass, I-300, VU, 230V. . Lift System: Elec. Heating Bath: w/o SJ 29/32 Glass set: w/o int.: I -300, valve unit 1 * 1 items
Catalog # BUCH11R300201N041
Supplier: BUCHI
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection