ContactUsLinkComponent
Rotavapor R-300 System B-305, SJ24/40, V, P+G, I-300, V-300, 230V. Inc. V-300 pump, I-300 int. w/o chiller. Lift System: Manual Heat ing Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: Vertical (V) P+G coating, Woulff bottle 1 * 1 items
Catalog # BUCH11SR300152VS01
Supplier:  BUCHI
Rotavapor R-300 System B-305, SJ24/40, V, P+G, I-300, V-300, 230V. Inc. V-300 pump, I-300 int. w/o chiller. Lift System: Manual Heat ing Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: Vertical (V) P+G coating, Woulff bottle 1 * 1 items
Catalog # BUCH11SR300152VS01
Supplier:  BUCHI
Frequently Bought Together