ContactUsLinkComponent
Rotavapor R-300 B-305, no joint., V, P+G, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Vert ical (V) P+G coating, int.: I-300, valve unit 1 * 1 items
Artikelnr. BUCH11R300150V141
Supplier:  BUCHI
Rotavapor R-300 B-305, no joint., V, P+G, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Vert ical (V) P+G coating, int.: I-300, valve unit 1 * 1 items
Artikelnr. BUCH11R300150V141
Supplier:  BUCHI
Frequently Bought Together