ContactUsLinkComponent
Rotavapor R-300 no bath, no joint., V, P+G, I-300, 230V. . Lift System: Manual Heating Bath: w/o Glass set: Vertical (V) P+G coating , int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300100V121
Supplier:  BUCHI
Rotavapor R-300 no bath, no joint., V, P+G, I-300, 230V. . Lift System: Manual Heating Bath: w/o Glass set: Vertical (V) P+G coating , int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300100V121
Supplier:  BUCHI
Frequently Bought Together