ContactUsLinkComponent
Rotavapor R-300 B-305, no joint., C, I-300P, 230V. . Elec. lift, B-305 bath, Glass set: Cold trap, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300250C031
Supplier:  BUCHI
Rotavapor R-300 B-305, no joint., C, I-300P, 230V. . Elec. lift, B-305 bath, Glass set: Cold trap, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300250C031
Supplier:  BUCHI
Frequently Bought Together