Rotavapor R-300 B-301, SJ29/32, V, I-300P, VU, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Vertical (V) int.: I-300 Pro, va lve unit 1 * 1 items
Catalog # BUCH11R300211V051
Supplier: BUCHI
Rotavapor R-300 B-301, SJ29/32, V, I-300P, VU, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Vertical (V) int.: I-300 Pro, va lve unit 1 * 1 items
Catalog # BUCH11R300211V051
Supplier: BUCHI
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection