ContactUsLinkComponent
testo 312-4 - Differential pressure measuring instrument up to 200 hPa for carrying out all necessary tests on gas heaters and pipes as well as water pipes. Inherently safe thanks to the use of C-Flex hoses with high impermeability. Legally compliant 1 * 1 items
Catalog # TEST0632.0327
Supplier:  Testo
testo 312-4 - Differential pressure measuring instrument up to 200 hPa for carrying out all necessary tests on gas heaters and pipes as well as water pipes. Inherently safe thanks to the use of C-Flex hoses with high impermeability. Legally compliant 1 * 1 items
Catalog # TEST0632.0327
Supplier:  Testo