ContactUsLinkComponent
Basic set testo 312-4: Differential pressure gauge testo 312-4 Hose set for testo 312-4 Balloon pump with release screw Conical test plug 1/2 Conical test plug 3/4 IR report printer Pressure set for gas pressure measurements on heating systems S 1 * 1 items
Catalog # TEST0563.1327
Supplier:  Testo
Basic set testo 312-4: Differential pressure gauge testo 312-4 Hose set for testo 312-4 Balloon pump with release screw Conical test plug 1/2 Conical test plug 3/4 IR report printer Pressure set for gas pressure measurements on heating systems S 1 * 1 items
Catalog # TEST0563.1327
Supplier:  Testo