Storage Sacks
Approx. 1250 bags/pack, 0,05 mm, 200×400 mm
LAMA117
LDPE, neutral storage bags.
- With bottom weld
- Can be used for foodstuffs
- Description:Approx. 1250 bags/pack
- Dimensions:200×400 mm
- Seq:03
- Thickness:0,05 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



